3-(Difluoromethyl)-5-methoxy-1H-indazole
Lieferant: Apollo Scientific
PC520139-1G
APOSPC520139-1GEA
2740
EUR
APOSPC520139-1G
3-(Difluoromethyl)-5-methoxy-1H-indazole
3-(Difluoromethyl)-5-methoxy-1H-indazole
Formel:
C₉H₈F₂N₂O Molecular Weight: 198,169 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767622 CAS-Nummer: 2149597-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...