5-Bromo-3-(difluoromethyl)-1H-indazole

Lieferant: Apollo Scientific

PC520142-1G
APOSPC520142-1GEA 1770 EUR
APOSPC520142-1G
5-Bromo-3-(difluoromethyl)-1H-indazole
5-Bromo-3-(difluoromethyl)-1H-indazole
Formel: C₈H₅BrF₂N₂
Molecular Weight: 247.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767625
CAS-Nummer: 1782324-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR