[3-(Difluoromethyl)-1H-indazol-5-yl]azinate

Lieferant: Apollo Scientific

PC520144-1G
APOSPC520144-1GEA 2280 EUR
APOSPC520144-1G
[3-(Difluoromethyl)-1H-indazol-5-yl]azinate
[3-(Difluoromethyl)-1H-indazol-5-yl]azinate
Formel: C₈H₅F₂N₃O₂
Molecular Weight: 213,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496615
CAS-Nummer: 2020068-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR