3-Bromo-5-(difluoromethyl)-1H-indazole
Lieferant: Apollo Scientific
PC520145-1G
APOSPC520145-1GEA
1770
EUR
APOSPC520145-1G
3-Bromo-5-(difluoromethyl)-1H-indazole
3-Bromo-5-(difluoromethyl)-1H-indazole
Formel:
C₈H₅BrF₂N₂ Molecular Weight: 247,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767627 CAS-Nummer: 2090721-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...