6-Bromo-3-(difluoromethyl)-7-methyl-imidazo[1,2-a]pyridine

Lieferant: Apollo Scientific

PC520152-1G
APOSPC520152-1GEA 3190 EUR
APOSPC520152-1G
6-Bromo-3-(difluoromethyl)-7-methyl-imidazo[1,2-a]pyridine
6-Bromo-3-(difluoromethyl)-7-methyl-imidazo[1,2-a]pyridine
Formel: C₉H₇BrF₂N₂
Molecular Weight: 261,066 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767633
CAS-Nummer: 2092048-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR