8-Bromo-3-(difluoromethyl)-6-methyl-1,8a-dihydroimidazo[1,2-a]pyridine

Lieferant: Apollo Scientific

PC520153-1G
APOSPC520153-1GEA 3190 EUR
APOSPC520153-1G
8-Bromo-3-(difluoromethyl)-6-methyl-1,8a-dihydroimidazo[1,2-a]pyridine
8-Bromo-3-(difluoromethyl)-6-methyl-1,8a-dihydroimidazo[1,2-a]pyridine
Formel: C₉H₉BrF₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29767634
CAS-Nummer: 2090237-67-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR