6-Bromo-3-(difluoromethyl)imidazo[1,2-a]pyridine

Lieferant: Apollo Scientific

PC520156-1G
APOSPC520156-1GEA 3190 EUR
APOSPC520156-1G
6-Bromo-3-(difluoromethyl)imidazo[1,2-a]pyridine
6-Bromo-3-(difluoromethyl)imidazo[1,2-a]pyridine
Formel: C₈H₅BrF₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29767636
CAS-Nummer: 2092522-63-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR