3-(Difluoromethyl)imidazo[1,2-a]pyridin-8-ol

Lieferant: Apollo Scientific

PC520157-1G
APOSPC520157-1GEA 3190 EUR
APOSPC520157-1G
3-(Difluoromethyl)imidazo[1,2-a]pyridin-8-ol
3-(Difluoromethyl)imidazo[1,2-a]pyridin-8-ol
Formel: C₈H₆F₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD29767637
CAS-Nummer: 2091580-80-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR