8-Bromo-6-chloro-3-(difluoromethyl)imidazo[1,2-a]pyridine
Lieferant: Apollo Scientific
PC520158-1G
APOSPC520158-1GEA
3190
EUR
APOSPC520158-1G
8-Bromo-6-chloro-3-(difluoromethyl)imidazo[1,2-a]pyridine
8-Bromo-6-chloro-3-(difluoromethyl)imidazo[1,2-a]pyridine
Formel:
C₈H₄BrClF₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767638 CAS-Nummer: 2092851-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...