6-Bromo-3-(difluoromethyl)imidazo[1,2-a]pyrazine

Lieferant: Apollo Scientific

PC520166-1G
APOSPC520166-1GEA 3190 EUR
APOSPC520166-1G
6-Bromo-3-(difluoromethyl)imidazo[1,2-a]pyrazine
6-Bromo-3-(difluoromethyl)imidazo[1,2-a]pyrazine
Formel: C₇H₄BrF₂N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD29767646
CAS-Nummer: 2092627-80-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR