4-Chloro-5-(difluoromethyl)-7H-pyrrolo[2,3-d]pyrimidine

Lieferant: Apollo Scientific

PC520168-1G
APOSPC520168-1GEA 4320 EUR
APOSPC520168-1G
4-Chloro-5-(difluoromethyl)-7H-pyrrolo[2,3-d]pyrimidine
4-Chloro-5-(difluoromethyl)-7H-pyrrolo[2,3-d]pyrimidine
Formel: C₇H₄ClF₂N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD29767648
CAS-Nummer: 2090261-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR