2-Bromo-7-(difluoromethyl)-5H-pyrrolo[2,3-b]pyrazine
Lieferant: Apollo Scientific
PC520169-1G
APOSPC520169-1GEA
2740
EUR
APOSPC520169-1G
2-Bromo-7-(difluoromethyl)-5H-pyrrolo[2,3-b]pyrazine
2-Bromo-7-(difluoromethyl)-5H-pyrrolo[2,3-b]pyrazine
Formel:
C₇H₄BrF₂N₃ Molecular Weight: 248,028 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767649 CAS-Nummer: 2092061-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...