Methyl 5-(difluoromethyl)-1H-pyrrole-2-carboxylate

Lieferant: Apollo Scientific

PC520170-1G
APOSPC520170-1GEA 1770 EUR
APOSPC520170-1G
Methyl 5-(difluoromethyl)-1H-pyrrole-2-carboxylate
Methyl 5-(difluoromethyl)-1H-pyrrole-2-carboxylate
Formel: C₇H₇F₂NO₂
Molecular Weight: 175,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767650
CAS-Nummer: 2112952-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR