2-(Difluoromethyl)-4-iodo-1H-pyrrole

Lieferant: Apollo Scientific

PC520176-1G
APOSPC520176-1GEA 2740 EUR
APOSPC520176-1G
2-(Difluoromethyl)-4-iodo-1H-pyrrole
2-(Difluoromethyl)-4-iodo-1H-pyrrole
Formel: C₅H₄F₂IN
Molecular Weight: 242,993 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767656
CAS-Nummer: 2091786-73-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR