4-(Difluoromethyl)-2-methyl-1H-imidazole

Lieferant: Apollo Scientific

PC520178-1G
APOSPC520178-1GEA 912 EUR
APOSPC520178-1G
4-(Difluoromethyl)-2-methyl-1H-imidazole
4-(Difluoromethyl)-2-methyl-1H-imidazole
Formel: C₅H₆F₂N₂
Molecular Weight: 132,111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767658
CAS-Nummer: 1684423-89-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR