5-(Difluoromethyl)oxazole

Lieferant: Apollo Scientific

PC520184-1G
APOSPC520184-1GEA 2280 EUR
APOSPC520184-1G
5-(Difluoromethyl)oxazole
5-(Difluoromethyl)oxazole
Formel: C₄H₃F₂NO
Molecular Weight: 119,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767664
CAS-Nummer: 2022038-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR