3-Bromo-2-chloro-5-(difluoromethyl)thiophene

Lieferant: Apollo Scientific

PC520188-1G
APOSPC520188-1GEA 2280 EUR
APOSPC520188-1G
3-Bromo-2-chloro-5-(difluoromethyl)thiophene
3-Bromo-2-chloro-5-(difluoromethyl)thiophene
Formel: C₅H₂BrClF₂S
Molecular Weight: 247,488 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767669
CAS-Nummer: 2091608-75-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR