4-Chloro-6-(difluoromethyl)thieno[3,2-d]pyrimidine

Lieferant: Apollo Scientific

PC520189-1G
APOSPC520189-1GEA 2740 EUR
APOSPC520189-1G
4-Chloro-6-(difluoromethyl)thieno[3,2-d]pyrimidine
4-Chloro-6-(difluoromethyl)thieno[3,2-d]pyrimidine
Formel: C₇H₃ClF₂N₂S
Molecular Weight: 220,627 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767670
CAS-Nummer: 2092627-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR