6-[5-(Difluoromethyl)-3-thienyl]pyrimidine-2,4-diol

Lieferant: Apollo Scientific

PC520198-1G
APOSPC520198-1GEA 2740 EUR
APOSPC520198-1G
6-[5-(Difluoromethyl)-3-thienyl]pyrimidine-2,4-diol
6-[5-(Difluoromethyl)-3-thienyl]pyrimidine-2,4-diol
Formel: C₉H₆F₂N₂O₂S
Molecular Weight: 244,218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767679
CAS-Nummer: 2092061-86-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR