2-(Difluoromethyl)-3-phenoxy-thiophene

Lieferant: Apollo Scientific

PC520200-1G
APOSPC520200-1GEA 1370 EUR
APOSPC520200-1G
2-(Difluoromethyl)-3-phenoxy-thiophene
2-(Difluoromethyl)-3-phenoxy-thiophene
Formel: C₁₁H₈F₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD29767681
CAS-Nummer: 2149598-79-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR