4-[5-(Difluoromethyl)-2-thienyl]-3-fluoro-phenol
Lieferant: Apollo Scientific
PC520209-1G
APOSPC520209-1GEA
2280
EUR
APOSPC520209-1G
4-[5-(Difluoromethyl)-2-thienyl]-3-fluoro-phenol
4-[5-(Difluoromethyl)-2-thienyl]-3-fluoro-phenol
Formel:
C₁₁H₇F₃OS Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767689 CAS-Nummer: 2090262-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...