5-[5-(Difluoromethyl)-2-thienyl]pyridin-2-ol
Lieferant: Apollo Scientific
PC520213-1G
APOSPC520213-1GEA
2740
EUR
APOSPC520213-1G
5-[5-(Difluoromethyl)-2-thienyl]pyridin-2-ol
5-[5-(Difluoromethyl)-2-thienyl]pyridin-2-ol
Formel:
C₁₀H₇F₂NOS Molecular Weight: 227,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767693 CAS-Nummer: 2092628-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...