5-(Difluoromethyl)-4-methyl-thiophene-2-carboxylic acid

Lieferant: Apollo Scientific

PC520214-1G
APOSPC520214-1GEA 2740 EUR
APOSPC520214-1G
5-(Difluoromethyl)-4-methyl-thiophene-2-carboxylic acid
5-(Difluoromethyl)-4-methyl-thiophene-2-carboxylic acid
Formel: C₇H₆F₂O₂S
Molecular Weight: 192,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767694
CAS-Nummer: 2092062-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR