3-Bromo-5-(difluoromethyl)-2-methyl-thiophene
Lieferant: Apollo Scientific
PC520223-1G
APOSPC520223-1GEA
2280
EUR
APOSPC520223-1G
3-Bromo-5-(difluoromethyl)-2-methyl-thiophene
3-Bromo-5-(difluoromethyl)-2-methyl-thiophene
Formel:
C₆H₅BrF₂S Molecular Weight: 227,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767703 CAS-Nummer: 2091602-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...