5-[5-(Difluoromethyl)-2-thienyl]pyridin-2-amine

Lieferant: Apollo Scientific

PC520226-1G
APOSPC520226-1GEA 3190 EUR
APOSPC520226-1G
5-[5-(Difluoromethyl)-2-thienyl]pyridin-2-amine
5-[5-(Difluoromethyl)-2-thienyl]pyridin-2-amine
Formel: C₁₀H₈F₂N₂S
Molecular Weight: 226,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767706
CAS-Nummer: 2090989-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR