4-[5-(Difluoromethyl)-2-thienyl]-2-fluoro-phenol

Lieferant: Apollo Scientific

PC520231-1G
APOSPC520231-1GEA 2280 EUR
APOSPC520231-1G
4-[5-(Difluoromethyl)-2-thienyl]-2-fluoro-phenol
4-[5-(Difluoromethyl)-2-thienyl]-2-fluoro-phenol
Formel: C₁₁H₇F₃OS
Lagertemperatur: Raumtemperatur
MDL: MFCD29767711
CAS-Nummer: 2092285-71-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR