5-(Difluoromethyl)thiophene-2-carbonitrile

Lieferant: Apollo Scientific

PC520235-1G
APOSPC520235-1GEA 1770 EUR
APOSPC520235-1G
5-(Difluoromethyl)thiophene-2-carbonitrile
5-(Difluoromethyl)thiophene-2-carbonitrile
Formel: C₆H₃F₂NS
Molecular Weight: 159,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767715
CAS-Nummer: 2112657-85-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR