5-[5-(Difluoromethyl)-2-thienyl]pyridin-3-ol
Lieferant: Apollo Scientific
PC520237-1G
APOSPC520237-1GEA
2280
EUR
APOSPC520237-1G
5-[5-(Difluoromethyl)-2-thienyl]pyridin-3-ol
5-[5-(Difluoromethyl)-2-thienyl]pyridin-3-ol
Formel:
C₁₀H₇F₂NOS Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767717 CAS-Nummer: 2090725-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...