2-(Difluoromethyl)-5-iodo-thiophene

Lieferant: Apollo Scientific

PC520242-1G
APOSPC520242-1GEA 2280 EUR
APOSPC520242-1G
2-(Difluoromethyl)-5-iodo-thiophene
2-(Difluoromethyl)-5-iodo-thiophene
Formel: C₅H₃F₂IS
Molecular Weight: 260,044 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767721
CAS-Nummer: 2092628-46-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR