3-[5-(Difluoromethyl)-2-thienyl]pyridin-2-ol

Lieferant: Apollo Scientific

PC520245-1G
APOSPC520245-1GEA 2280 EUR
APOSPC520245-1G
3-[5-(Difluoromethyl)-2-thienyl]pyridin-2-ol
3-[5-(Difluoromethyl)-2-thienyl]pyridin-2-ol
Formel: C₁₀H₇F₂NOS
Molecular Weight: 227,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767724
CAS-Nummer: 2091581-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR