Ethyl 5-(difluoromEthyl)thiophene-3-carboxylate
Lieferant: Apollo Scientific
PC520250-1G
APOSPC520250-1GEA
2280
EUR
APOSPC520250-1G
Ethyl 5-(difluoromEthyl)thiophene-3-carboxylate
Ethyl 5-(difluoromEthyl)thiophene-3-carboxylate
Formel:
C₈H₈F₂O₂S Molecular Weight: 206.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767729 CAS-Nummer: 189331-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...