4-Bromo-2-(difluoromethyl)benzothiophene

Lieferant: Apollo Scientific

PC520255-1G
APOSPC520255-1GEA 2270 EUR
APOSPC520255-1G
4-Bromo-2-(difluoromethyl)benzothiophene
4-Bromo-2-(difluoromethyl)benzothiophene
Formel: C₉H₅BrF₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD29767735
CAS-Nummer: 1785254-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR