2-(Difluoromethyl)-3-methoxy-thiophene

Lieferant: Apollo Scientific

PC520256-1G
APOSPC520256-1GEA 1830 EUR
APOSPC520256-1G
2-(Difluoromethyl)-3-methoxy-thiophene
2-(Difluoromethyl)-3-methoxy-thiophene
Formel: C₆H₆F₂OS
Molecular Weight: 164,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767736
CAS-Nummer: 2149591-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR