2,5-Dibromo-3-(difluoromethyl)thiophene

Lieferant: Apollo Scientific

PC520257-1G
APOSPC520257-1GEA 0 EUR
APOSPC520257-1G
2,5-Dibromo-3-(difluoromethyl)thiophene
2,5-Dibromo-3-(difluoromethyl)thiophene
Formel: C₅H₂Br₂F₂S
Molecular Weight: 291,939 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767737
CAS-Nummer: 1244041-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR