2-Bromo-5-chloro-3-(difluoromethyl)thiophene
Lieferant: Apollo Scientific
PC520258-1G
APOSPC520258-1GEA
2280
EUR
APOSPC520258-1G
2-Bromo-5-chloro-3-(difluoromethyl)thiophene
2-Bromo-5-chloro-3-(difluoromethyl)thiophene
Formel:
C₅H₂BrClF₂S Molecular Weight: 247,488 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767738 CAS-Nummer: 2090725-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...