2-Bromo-3-(difluoromethyl)thiophene
Lieferant: Apollo Scientific
PC520259-1G
APOSPC520259-1GEA
2060
EUR
APOSPC520259-1G
2-Bromo-3-(difluoromethyl)thiophene
2-Bromo-3-(difluoromethyl)thiophene
Formel:
C₅H₃BrF₂S Molecular Weight: 213,043 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767739 CAS-Nummer: 2092062-85-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...