2-(2-Bromophenyl)-4-(difluoromethyl)thiazole
Lieferant: Apollo Scientific
PC520265-1G
APOSPC520265-1GEA
1830
EUR
APOSPC520265-1G
2-(2-Bromophenyl)-4-(difluoromethyl)thiazole
2-(2-Bromophenyl)-4-(difluoromethyl)thiazole
Formel:
C₁₀H₆BrF₂NS Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767745 CAS-Nummer: 2091121-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...