tert-Butyl N-[[4-(difluoromethyl)thiazol-2-yl]methyl]carbamate

Lieferant: Apollo Scientific

PC520267-1G
APOSPC520267-1GEA 2740 EUR
APOSPC520267-1G
tert-Butyl N-[[4-(difluoromethyl)thiazol-2-yl]methyl]carbamate
tert-Butyl N-[[4-(difluoromethyl)thiazol-2-yl]methyl]carbamate
Formel: C₁₀H₁₄F₂N₂O₂S
Molecular Weight: 264,292 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767748
CAS-Nummer: 165667-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR