4-(Difluoromethyl)-2-methyl-thiazole

Lieferant: Apollo Scientific

PC520268-1G
APOSPC520268-1GEA 1430 EUR
APOSPC520268-1G
4-(Difluoromethyl)-2-methyl-thiazole
4-(Difluoromethyl)-2-methyl-thiazole
Formel: C₅H₅F₂NS
Molecular Weight: 149,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767749
CAS-Nummer: 1849218-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR