4-Bromo-5-(difluoromethyl)thiazole
Lieferant: Apollo Scientific
PC520275-1G
APOSPC520275-1GEA
2280
EUR
APOSPC520275-1G
4-Bromo-5-(difluoromethyl)thiazole
4-Bromo-5-(difluoromethyl)thiazole
Formel:
C₄H₂BrF₂NS Molecular Weight: 214.031 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767756 CAS-Nummer: 2090263-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...