2-(4-Bromophenyl)-5-(difluoromethyl)thiazole

Lieferant: Apollo Scientific

PC520278-1G
APOSPC520278-1GEA 2280 EUR
APOSPC520278-1G
2-(4-Bromophenyl)-5-(difluoromethyl)thiazole
2-(4-Bromophenyl)-5-(difluoromethyl)thiazole
Formel: C₁₀H₆BrF₂NS
Molecular Weight: 290,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767759
CAS-Nummer: 2090725-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR