4-Chloro-5-(difluoromethyl)thiazol-2-amine

Lieferant: Apollo Scientific
Achtung

PC520279-1G
APOSPC520279-1GEA 2280 EUR
APOSPC520279-1G
4-Chloro-5-(difluoromethyl)thiazol-2-amine
4-Chloro-5-(difluoromethyl)thiazol-2-amine
Formel: C₄H₃ClF₂N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD29767760
CAS-Nummer: 2092062-99-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR