1,3-Dibromo-5-(difluoromethyl)-2-methoxy-benzene

Lieferant: Apollo Scientific

PC520289-1G
APOSPC520289-1GEA 1710 EUR
APOSPC520289-1G
1,3-Dibromo-5-(difluoromethyl)-2-methoxy-benzene
1,3-Dibromo-5-(difluoromethyl)-2-methoxy-benzene
Formel: C₈H₆Br₂F₂O
Molecular Weight: 315,937 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496609
CAS-Nummer: 2149591-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR