tert-Butyl 4-(difluoromethyl)benzoate

Lieferant: Apollo Scientific

PC520292-1G
APOSPC520292-1GEA 2060 EUR
APOSPC520292-1G
tert-Butyl 4-(difluoromethyl)benzoate
tert-Butyl 4-(difluoromethyl)benzoate
Formel: C₁₂H₁₄F₂O₂
Molecular Weight: 228,235 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496611
CAS-Nummer: 1643376-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR