1-benzyloxy-2-(difluoromethyl)benzene
Lieferant: Apollo Scientific
PC520293-1G
APOSPC520293-1GEA
458
EUR
APOSPC520293-1G
1-benzyloxy-2-(difluoromethyl)benzene
1-benzyloxy-2-(difluoromethyl)benzene
Formel:
C₁₄H₁₂F₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767774 CAS-Nummer: 915799-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...