4-(Difluoromethyl)-2,6-Difluoro-benzene-1,3-diol

Lieferant: Apollo Scientific

PC520294-1G
APOSPC520294-1GEA 1710 EUR
APOSPC520294-1G
4-(Difluoromethyl)-2,6-Difluoro-benzene-1,3-diol
4-(Difluoromethyl)-2,6-Difluoro-benzene-1,3-diol
Formel: C₇H₄F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD30496612
CAS-Nummer: 2092183-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR