Methyl 2-(difluoromethyl)-3,5-dimethoxy-benzoate

Lieferant: Apollo Scientific

PC520295-1G
APOSPC520295-1GEA 912 EUR
APOSPC520295-1G
Methyl 2-(difluoromethyl)-3,5-dimethoxy-benzoate
Methyl 2-(difluoromethyl)-3,5-dimethoxy-benzoate
Formel: C₁₁H₁₂F₂O₄
Molecular Weight: 246,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496613
CAS-Nummer: 2149590-59-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR