2-Bromo-4-(difluoromethyl)-6-fluoro-aniline

Lieferant: Apollo Scientific

PC520298-1G
APOSPC520298-1GEA 1490 EUR
APOSPC520298-1G
2-Bromo-4-(difluoromethyl)-6-fluoro-aniline
2-Bromo-4-(difluoromethyl)-6-fluoro-aniline
Formel: C₇H₅BrF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD30496617
CAS-Nummer: 2092183-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR