4-(Difluoromethyl)-3,5-dimethoxy-phenol

Lieferant: Apollo Scientific

PC520300-1G
APOSPC520300-1GEA 1200 EUR
APOSPC520300-1G
4-(Difluoromethyl)-3,5-dimethoxy-phenol
4-(Difluoromethyl)-3,5-dimethoxy-phenol
Formel: C₉H₁₀F₂O₃
Molecular Weight: 204,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496619
CAS-Nummer: 2092380-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR