[4-(Difluoromethyl)-2,6-dimethoxy-phenyl] acetate
Lieferant: Apollo Scientific
PC520302-1G
APOSPC520302-1GEA
1200
EUR
APOSPC520302-1G
[4-(Difluoromethyl)-2,6-dimethoxy-phenyl] acetate
[4-(Difluoromethyl)-2,6-dimethoxy-phenyl] acetate
Formel:
C₁₁H₁₂F₂O₄ Molecular Weight: 246,207 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30496621 CAS-Nummer: 2149601-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...